accessibility menu, dialog, popup

UserName

CAS RN 756525-90-3

H3COOOOOOOOOONOO

CAS RN 756525-90-3

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2,5,8,11,14,17,20,23-octaoxahexacosan-26-oate
Synonyms: 2,5-dioxopyrrolidin-1-yl 2,5,8,11,14,17,20,23-octaoxahexacosan-26-oate
Molecular Weight (g/mol): 509.55
Molecular Formula: C22H39NO12
InChi Key: IBZNTYBFTKFSMU-UHFFFAOYSA-N
SMILES: COCCOCCOCCOCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results