Learn More
Methyl-PEG8-NHS Ester, TCI America™
Supplier:  TCI America M218725MG
Catalog No. M218725MG
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
Methyl-PEG8-NHS Ester
Colorless
MFCD13184969
N-Succinimidyl 4,7,10,13,16,19,22,25-Octaoxahexacosanoate, 4,7,10,13,16,19,22,25-Octaoxahexacosanoic Acid N-Succinimidyl Ester, mPEG8-NHS Ester
COCCOCCOCCOCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O
509.549
509.55
756525-90-3
C22H39NO12
25 mg
IBZNTYBFTKFSMU-UHFFFAOYSA-N
(2,5-dioxopyrrolidin-1-yl) 3-[2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]propanoate
51340981
Liquid
Chemical Identifiers
756525-90-3
509.549
IBZNTYBFTKFSMU-UHFFFAOYSA-N
51340981
COCCOCCOCCOCCOCCOCCOCCOCCC(=O)ON1C(=O)CCC1=O
C22H39NO12
MFCD13184969
N-Succinimidyl 4,7,10,13,16,19,22,25-Octaoxahexacosanoate, 4,7,10,13,16,19,22,25-Octaoxahexacosanoic Acid N-Succinimidyl Ester, mPEG8-NHS Ester
(2,5-dioxopyrrolidin-1-yl) 3-[2-[2-[2-[2-[2-[2-(2-methoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]propanoate
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
TSCA : No
Recommended Storage : Freezer