CAS RN 75400-67-8
IUPAC Name:
tert-butyl 1H-indole-1-carboxylate
Synonyms:
tert-butyl indole-1-carboxylate
Molecular Weight (g/mol):
217.27
Molecular Formula:
C13H15NO2
InChi Key:
OWPIFQXNMLDXKW-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)N1C=CC2=CC=CC=C12
1
–
1
of
1
results
1
Promotions Available
1-Boc-indole, 97%
1-Boc-indole, 97%
CAS: 75400-67-8 Molecular Formula: C13H15NO2 Molecular Weight (g/mol): 217.27 MDL Number: MFCD02093939 InChI Key: OWPIFQXNMLDXKW-UHFFFAOYSA-N Synonym: 1-boc-indole,tert-butyl 1h-indole-1-carboxylate,n-boc-indole,tert-butyl 1-indolecarboxylate,1h-indole-1-carboxylic acid, 1,1-dimethylethyl ester,1-tert-butoxycarbonyl indole,n-boc indole,pubchem7228,acmc-1bjgd,1-tert-butoxycarbonylindole PubChem CID: 3532980 SMILES: CC(C)(C)OC(=O)N1C=CC2=CC=CC=C12