accessibility menu, dialog, popup

UserName

H2NO(S)H2N

CAS RN 7531-52-4

IUPAC Name: (2S)-2-carbamoylpyrrolidin-1-ium
Synonyms: L-prolinamide
Molecular Weight (g/mol): 115.16
Molecular Formula: C5H11N2O
InChi Key: VLJNHYLEOZPXFW-BYPYZUCNSA-O
SMILES: NC(=O)[C@@H]1CCC[NH2+]1
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)