missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Prolinamide, 98%

Catalog No. AAL2041706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2041706 5 g
AAL2041703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2041706 Supplier Thermo Scientific Chemicals Supplier No. L2041706

CAS: 7531-52-4 | C5H11N2O | 115.16 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also it is a efficient organocatalysts for aldol reactions on water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff. Also it is a efficient organocatalysts for aldol reactions on water.

Solubility
Soluble in water and ethanol (50 mg/ml).

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 7531-52-4
Molecular Formula C5H11N2O
Molecular Weight (g/mol) 115.16
MDL Number MFCD00005253
InChI Key VLJNHYLEOZPXFW-BYPYZUCNSA-O
Synonym l-prolinamide, prolinamide, h-pro-nh2, s-pyrrolidine-2-carboxamide, s-prolinamide, 2s-pyrrolidine-2-carboxamide, l-prolineamide, l-proline amide, l---prolinamide, s-2-pyrrolidinecarboxamide
PubChem CID 111306
ChEBI CHEBI:21374
SMILES NC(=O)[C@@H]1CCC[NH2+]1

Specifications

Melting Point 95°C to 104°C
Quantity 5 g
Beilstein 80807
Solubility Information Soluble in water and ethanol (50mg/ml).
Optical Rotation −105° (c=2 in Ethanol)
IUPAC Name (2S)-2-carbamoylpyrrolidin-1-ium
Formula Weight 114.15
Percent Purity 99%
Chemical Name or Material L-(-)-Prolinamide
EINECSNumber 231-397-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only