accessibility menu, dialog, popup

UserName

CH3H3COOOO(E/Z)

CAS RN 7529-87-5

IUPAC Name: 4-oxo-4-(propan-2-yloxy)but-2-enoate
Synonyms: 4-isopropoxy-4-oxobut-2-enoate
Molecular Weight (g/mol): 157.15
Molecular Formula: C7H9O4
InChi Key: FWUIHQFQLSWYED-UHFFFAOYSA-M
SMILES: CC(C)OC(=O)C=CC([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results