missing translation for 'onlineSavingsMsg'
Learn More

Monoisopropyl Fumarate 97.0+%, TCI America™

Catalog No. M178425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M178425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M178425G Supplier TCI America Supplier No. M178425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7529-87-5
Molecular Formula C7H9O4-
Molecular Weight (g/mol) 157.145
InChI Key FWUIHQFQLSWYED-ONEGZZNKSA-M
Synonym Fumaric Acid Monoisopropyl Ester, Isopropyl Hydrogen Fumarate
PubChem CID 21889704
IUPAC Name (E)-4-oxo-4-propan-2-yloxybut-2-enoate
SMILES CC(C)OC(=O)C=CC(=O)[O-]

Specifications

Melting Point 48°C
Color White
Quantity 25 g
Formula Weight 158.15
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Monoisopropyl Fumarate
EINECSNumber (2)-1142
TSCA No
Recommended Storage Refrigerator