man

Test

2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 7512-43-8

CH3OONH2SH3CHCl

CAS RN 7512-43-8

IUPAC Name: hydrogen ethyl 2-amino-4-(methylsulfanyl)butanoate chloride
Synonyms: hydrogen ethyl 2-amino-4-(methylsulfanyl)butanoate chloride
Molecular Weight (g/mol): 213.72
Molecular Formula: C7H16ClNO2S
InChi Key: KPWCQEUBJAIORR-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCOC(=O)C(N)CCSC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)