accessibility menu, dialog, popup

UserName

CAS RN 75-99-0

CH3ClClOHO

CAS RN 75-99-0

IUPAC Name: 2,2-dichloropropanoic acid
Synonyms: dalapon
Molecular Weight (g/mol): 142.96
Molecular Formula: C3H4Cl2O2
InChi Key: NDUPDOJHUQKPAG-UHFFFAOYSA-N
SMILES: CC(Cl)(Cl)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results