missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Dichloropropionic Acid 95.0+%, TCI America™
Supplier: TCI America D13341G
Specifications
| 2,2-Dichloropropionic Acid | |
| Yellow | |
| C3H4Cl2O2 | |
| 1 g | |
| NDUPDOJHUQKPAG-UHFFFAOYSA-N | |
| 2,2-dichloropropanoic acid | |
| 6418 | |
| 142.96 | |
| Liquid |
| 75-99-0 | |
| 99°C | |
| MFCD00004178 | |
| 2,2-dichloropropionic acid, dalapon, kenapon, liropon, proprop, alatex, unipon, sys-omnidel, d-granulat, dawpon-rae | |
| CC(C(=O)O)(Cl)Cl | |
| 142.963 | |
| CHEBI:81859 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 75-99-0 | |
| 142.963 | |
| NDUPDOJHUQKPAG-UHFFFAOYSA-N | |
| 6418 | |
| 2,2-dichloropropanoic acid |
| C3H4Cl2O2 | |
| MFCD00004178 | |
| 2,2-dichloropropionic acid, dalapon, kenapon, liropon, proprop, alatex, unipon, sys-omnidel, d-granulat, dawpon-rae | |
| CHEBI:81859 | |
| CC(C(=O)O)(Cl)Cl |
Safety and Handling
EINECSNumber : (2)-3913
RTECSNumber : UF0690000
TSCA : Yes