accessibility menu, dialog, popup

UserName

SCS

CAS RN 75-15-0

IUPAC Name: methanedithione
Synonyms: carbon disulfide carbon disulphide
Molecular Weight (g/mol): 76.13
Molecular Formula: CS2
InChi Key: QGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILES: S=C=S
Molecular Weight (g/mol) 
  • (2)
  • (27)
Quantity 
  • (7)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (9)
  • (1)
  • (1)
Grade 
  • (8)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (10)
  • (4)
  • (1)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (4)
  • (4)
  • (3)
  • (3)
  • (1)
  • (2)
  • (12)
  • (2)
  • (27)
  • (7)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (9)
  • (1)
  • (1)
  • (8)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (10)
  • (4)
  • (1)
  • (5)