missing translation for 'onlineSavingsMsg'
Learn More

Carbon Disulfide (Spectranalyzed™), Fisher Chemical

Catalog No. C573500
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
C573500 500 mL Amber Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C573500 Supplier Thermo Fisher Scientific Supplier No. FLC573500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use in ultraviolet and infrared spectroscopy

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point -111°C
Absorbance Pass Test
Color Yellow
Quantity 500 mL
Flash Point -30°C (-22°F)
Identification Pass Test
Infrared Spectrum Pass Test
Packaging Amber Glass
Physical Form Liquid
Residue after Evaporation 0.002% max.
Assay Percent Range ≥99.9 %
Vapor Pressure 297.5mmHg at 20°C
Viscosity 0.363 cps at 20 deg C
Water 0.05% max.
Boiling Point 46°C
Formula Weight 76.14
Percent Purity ≥99.9%
Grade Also meets ACS specifications
Chemical Name or Material Carbon Disulfide
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Harmful by inhalation, in contact with skin and if swallowed. Keep container tightly closed. Ensure adequate ventilation. . . . . .

NFPA

Health:3
Flammability:4
Instability:0