missing translation for 'onlineSavingsMsg'
Learn More

Carbon Disulfide, MilliporeSigma™

Catalog No. M1022101000
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1022101000 1 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1022101000 Supplier Technidata Supplier No. 1.02210.1000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For spectroscopy Uvasol™

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point -111.6°C
Color Colorless
Density 1.26g/cm3 at 20°C
Quantity 1 L
Flash Point −30°C (−22°F)
Ignition Point 95°C (203°F)
Odor Charactristic
Packaging Glass Bottle
Physical Form Liquid
Vapor Pressure 398 hPa (20°C)
UN Number 1131
Boiling Point 46.5°C
Solubility Information Soluble in water: 2.1g/L at 20°C
Formula Weight 76.13
Chemical Name or Material Carbon Disulfide
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H315 Causes skin irritation.
Precautionary Statement H319: Causes serious eye irritation.
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.