accessibility menu, dialog, popup

UserName

Ho

CAS RN 7440-60-0

IUPAC Name: holmium
Synonyms: holmium|holmium atom
Molecular Weight (g/mol): 164.93
Molecular Formula: Ho
InChi Key: KJZYNXUDTRRSPN-UHFFFAOYSA-N
SMILES: [Ho]
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
Form 
  • (2)
  • (4)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (11)