missing translation for 'onlineSavingsMsg'
Learn More

Potassium standard solution, for AAS, 1 mg/ml K in 2-5% HNO3

Catalog No. AC196205000
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC196205000 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC196205000 Supplier Thermo Scientific Chemicals Supplier No. 196205000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-09-7 | K | 39.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for atomic adsorption spectroscopy

Specifications

Chemical Name or Material Potassium standard solution
Packaging Plastic bottle
Name Note 1 mg/ml K in 2% HNO3
CAS 7440-09-7, 7732-18-5
Physical Form Liquid
Molecular Formula K
Concentration 997 to 1003 μg/mL of K(1+) in 2% HNO3 (20°C)
MDL Number MFCD00133776
Color Colorless
Solubility Miscible with water
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
SMILES [K]
IUPAC Name potassium
Molecular Weight (g/mol) 39.10
Quantity 500 mL
PubChem CID 5462222
ChEBI CHEBI:26216
Formula Weight 39.1
Grade AAS
Show More Show Less

RUO – Research Use Only