CAS RN 74199-16-9
CAS RN 74199-16-9
IUPAC Name:
1,4,8,11-tetraazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadecane
Synonyms:
1,4,8,11-tetraazatetracyclo[6.6.2.0⁴,¹⁶.0¹¹,¹⁵]hexadecane
Molecular Weight (g/mol):
222.34
Molecular Formula:
C12H22N4
InChi Key:
CMHJBNKGPWROQM-UHFFFAOYSA-N
SMILES:
C1CN2CCN3CCCN4CCN(C1)C2C34
1
–
1
of
1
results
1
cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene 98.0+%, TCI America™
CAS: 74199-16-9 Molecular Formula: C12H22N4 Molecular Weight (g/mol): 222.336 MDL Number: MFCD04038934 InChI Key: CMHJBNKGPWROQM-UHFFFAOYSA-N Synonym: cyclan glyoxal, cis-ptap,cis-decahydro-1h,6h-3a,5a,8a,10a-tetraazapyrene,cis-glyoxal-cyclam,cyclam cis-glyoxal derivative,cis-1,4,8,11-perhydrotetra azapyrene,10bbeta,10cbeta-decahydro-1h,6h-3a,5a,8a,10a-tetraazapyrene,1,4,8,11-tetraazatetracyclo 6.6.2.0?, 1 ?.0 1 1 , 1 ? hexadecane,2,3,4,5,7,8,9,10,10b,10c-decahydro-1h,6h-3a,5a,8a,10a-tetraazapyrene,15s,16s-1,4,8,11-tetraazatetracyclo 6.6.2.0?, 1 ?.0 1 1 , 1 ? hexadecane PubChem CID: 434804 SMILES: C1CN2CCN3CCCN4C3C2N(C1)CC4