missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 83°C |
| Color | White |
| Quantity | 100 mg |
| Formula Weight | 222.34 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | cis-Decahydro-1H,6H-3a,5a,8a,10a-tetraazapyrene |
Safety and Handling
| TSCA | No |