accessibility menu, dialog, popup

UserName

CH2OO

CAS RN 7398-56-3

IUPAC Name: tricyclo[5.2.1.0²,⁶]decan-8-yl prop-2-enoate
Synonyms: tricyclo[5.2.1.0²,⁶]decan-8-yl prop-2-enoate
Molecular Weight (g/mol): 206.29
Molecular Formula: C13H18O2
InChi Key: BDYXPMNDZDGGTI-UHFFFAOYNA-N
SMILES: C=CC(=O)OC1CC2CC1C1CCCC21
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results