missing translation for 'onlineSavingsMsg'
Learn More

Dicyclopentanyl Acrylate (stabilized with MEHQ) 95.0+%, TCI America™

Catalog No. D3380500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D3380500G 500 g
D338025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D3380500G Supplier TCI America Supplier No. D3380500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7398-56-3
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD08276368
InChI Key BDYXPMNDZDGGTI-UHFFFAOYNA-N
Synonym Acrylic Acid Dicyclopentanyl Ester, Acrylic Acid Tetrahydrodicyclopentadienyl Ester, Tetrahydrodicyclopentadienyl Acrylate
PubChem CID 13001872
IUPAC Name tricyclo[5.2.1.0²,⁶]decan-8-yl prop-2-enoate
SMILES C=CC(=O)OC1CC2CC1C1CCCC21

Specifications

Color Colorless
Quantity 500 g
Formula Weight 206.29
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Dicyclopentanyl Acrylate (stabilized with MEHQ)
EINECSNumber (4)-1583
TSCA No
Recommended Storage Freezer