CAS RN 73276-70-7
CAS RN 73276-70-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
9-Ethylcarbazole-3-carboxaldehyde Diphenylhydrazone 98.0+%, TCI America™
CAS: 73276-70-7 Molecular Formula: C27H23N3 Molecular Weight (g/mol): 389.50 MDL Number: MFCD00144879 InChI Key: CEAPHJPESODIQL-VFCFBJKWSA-N Synonym: 3-(Diphenylhydrazonomethyl)-9-ethylcarbazole PubChem CID: 9577933 IUPAC Name: 3-[(E)-(2,2-diphenylhydrazin-1-ylidene)methyl]-9-ethyl-9H-carbazole SMILES: CCN1C2=C(C=CC=C2)C2=C1C=CC(\C=N\N(C1=CC=CC=C1)C1=CC=CC=C1)=C2