accessibility menu, dialog, popup

UserName

NBrBrBr

CAS RN 73087-83-9

IUPAC Name: 3,6-dibromo-9-(4-bromophenyl)-9H-carbazole
Synonyms: 3,6-dibromo-9-(4-bromophenyl)carbazole
Molecular Weight (g/mol): 480
Molecular Formula: C18H10Br3N
InChi Key: NRTDFHUSNYJENJ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results