missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-9-(4-bromophenyl)carbazole 98.0+%, TCI America™

Catalog No. D4563200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4563200MG 200 mg
D45631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4563200MG Supplier TCI America Supplier No. D4563200MG
Only null left

Chemical Identifiers

CAS 73087-83-9
Molecular Formula C18H10Br3N
Molecular Weight (g/mol) 480.00
MDL Number MFCD12024284
InChI Key NRTDFHUSNYJENJ-UHFFFAOYSA-N
PubChem CID 11059889
IUPAC Name 3,6-dibromo-9-(4-bromophenyl)-9H-carbazole
SMILES BrC1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2

Specifications

Melting Point 209°C
Color White-Yellow
Quantity 200 mg
Formula Weight 480.00
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-(4-bromophenyl)carbazole