accessibility menu, dialog, popup

UserName

CH3ONHOHH3CO

CAS RN 7298-67-1

IUPAC Name: N-(3-acetyl-4-hydroxyphenyl)acetamide
Synonyms: N-(3-acetyl-4-hydroxyphenyl)acetamide
Molecular Weight (g/mol): 193.2
Molecular Formula: C10H11NO3
InChi Key: DIQSYMRVTOVKQT-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(O)C(=C1)C(C)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)