missing translation for 'onlineSavingsMsg'
Learn More

5'-Acetamido-2'-hydroxyacetophenone 98.0+%, TCI America™

Catalog No. A26985G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26985G 5 g
A26981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A26985G Supplier TCI America Supplier No. A26985G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7298-67-1
Molecular Formula C10H11NO3
Molecular Weight (g/mol) 193.202
MDL Number MFCD00100490
InChI Key DIQSYMRVTOVKQT-UHFFFAOYSA-N
Synonym n-3-acetyl-4-hydroxyphenyl acetamide, n1-3-acetyl-4-hydroxyphenyl acetamide, n-3-acetyl-4-hydroxy-phenyl acetamide, 5'-acetamido-2'-hydroxyacetophenone, acetamide, n-3-acetyl-4-hydroxyphenyl, ksc380g7h, 3'-acetyl-4'-hydroxyacetanilide, 5-acetamido-2-hydroxyacetophenone, 5-acetamido-2-hydroxy-acetophenone, n-3-acetyl-4-hydroxyphenyl-acetamide
PubChem CID 81720
IUPAC Name N-(3-acetyl-4-hydroxyphenyl)acetamide
SMILES CC(=O)C1=C(C=CC(=C1)NC(=O)C)O

Specifications

Melting Point 168°C
Quantity 5 g
Formula Weight 193.20
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Acetamido-2′-hydroxyacetophenone