accessibility menu, dialog, popup

UserName

BOOCH3CH3H3CCH3BOOH3CH3CCH3H3C

CAS RN 728911-52-2

IUPAC Name: 4,4,5,5-tetramethyl-2-[7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9'-spirobi[fluoren]-2-yl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[7-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9'-spirobi[fluoren]-2-yl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 568.33
Molecular Formula: C37H38B2O4
InChi Key: GKPGYJFGTIZCRP-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)C1=C(C=C(C=C1)B1OC(C)(C)C(C)(C)O1)C21C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results