missing translation for 'onlineSavingsMsg'
Learn More

2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™

Catalog No. B56561G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B56561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B56561G Supplier TCI America Supplier No. B56561G

Chemical Identifiers

CAS 728911-52-2
Molecular Formula C37H38B2O4
Molecular Weight (g/mol) 568.327
InChI Key GKPGYJFGTIZCRP-UHFFFAOYSA-N
Synonym 2,2′-(9,9′-Spirobi[9H-fluorene]-2,7-diyl)bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolane), (9,9′-Spirobi[9H-fluorene]-2,7-diyl)bis(boronic Acid Pinacol Ester)
PubChem CID 20769414
IUPAC Name 4,4,5,5-tetramethyl-2-[7'-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9'-spirobi[fluorene]-2'-yl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)C4=C(C35C6=CC=CC=C6C7=CC=CC=C57)C=C(C=C4)B8OC(C(O8)(C)C)(C)C

Specifications

Melting Point 332°C
Color White-Yellow
Quantity 1 g
Formula Weight 568.33
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9,9′-spirobi[9H-fluorene]