accessibility menu, dialog, popup

UserName

CH3H3COBCH2OH3CH3C

CAS RN 72824-04-5

IUPAC Name: 4,4,5,5-tetramethyl-2-(prop-2-en-1-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(prop-2-en-1-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 168.04
Molecular Formula: C9H17BO2
InChi Key: YMHIEPNFCBNQQU-UHFFFAOYSA-N
SMILES: CC1(C)OB(CC=C)OC1(C)C
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (4)