missing translation for 'onlineSavingsMsg'
Learn More

2-Allyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane (stabilized with Phenothiazine) 96.0+%, TCI America™

Catalog No. A21575G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21575G 5 g
A21571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21575G Supplier TCI America Supplier No. A21575G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72824-04-5
Molecular Formula C9H17BO2
Molecular Weight (g/mol) 168.04
MDL Number MFCD00013347
InChI Key YMHIEPNFCBNQQU-UHFFFAOYSA-N
Synonym allylboronic acid pinacol ester, 2-allyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, allylboronic acid, pinacol cyclic ester, 4,4,5,5-tetramethyl-2-prop-2-en-1-yl-1,3,2-dioxaborolane, allylboronic acid piracol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-propenyl, 2-prop-2-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, allylboronicacidpinacolester, pinacol allylboronate, abape
PubChem CID 2763171
IUPAC Name 4,4,5,5-tetramethyl-2-(prop-2-en-1-yl)-1,3,2-dioxaborolane
SMILES CC1(C)OB(CC=C)OC1(C)C

Specifications

Color Yellow
Boiling Point 35°C
Quantity 5 g
UN Number 3272
Formula Weight 168.04
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Allyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane (stabilized with Phenothiazine)
TSCA No
Recommended Storage Freezer