accessibility menu, dialog, popup

UserName

H2N(R)NH2OHOHCl

CAS RN 7274-88-6

IUPAC Name: (2R)-2,6-diaminohexanoic acid hydrochloride
Synonyms: D-lysine hydrochloride
Molecular Weight (g/mol): 182.65
Molecular Formula: C6H15ClN2O2
InChi Key: BVHLGVCQOALMSV-XZNNNFJINA-N
SMILES: Cl.NCCCC[C@@H](N)C(O)=O
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)