missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Lysine Monohydrochloride 98.0+%, TCI America™

Catalog No. L01285G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01285G 5 g
L012825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01285G Supplier TCI America Supplier No. L01285G
Only null left

Chemical Identifiers

CAS 7274-88-6
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012920
InChI Key BVHLGVCQOALMSV-XZNNNFJINA-N
Synonym d-lysine hydrochloride, d-lysine monohydrochloride, d-lysine hcl, d-lysine, monohydrochloride, h-d-lys-oh.hcl, d-lysine, hydrochloride, unii-kk611jde1n, lysine, monohydrochloride, d, r-2,6-diaminohexanoic acid hydrochloride, lysine, hydrochloride, d
PubChem CID 81691
ChEBI CHEBI:53558
IUPAC Name (2R)-2,6-diaminohexanoic acid hydrochloride
SMILES Cl.NCCCC[C@@H](N)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 182.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material D-(-)-Lysine Monohydrochloride
EINECSNumber (2)-3200
RTECSNumber OL5632500
TSCA Yes