accessibility menu, dialog, popup

UserName

CAS RN 72679-02-8

IUPAC Name: 3-(benzoylsulfanyl)-2-methylpropanoic acid
Synonyms: 3-(benzoylsulfanyl)-2-methylpropanoic acid
Molecular Weight (g/mol): 224.27
Molecular Formula: C11H12O3S
InChi Key: BCAYPPFBOJCRPN-UHFFFAOYNA-N
SMILES: CC(CSC(=O)C1=CC=CC=C1)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results