CAS RN 72561-05-8
CAS RN 72561-05-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Endoproteinase Lys-C, Sequencing Grade, Lysobacter enzymogenes
Designed for protein sequencing or sequence verification, analysis of protein structural domains, and cleavage of fusion proteins