CAS RN 72548-79-9
IUPAC Name:
3-(pyrrolidin-1-yl)benzoic acid
Synonyms:
3-(pyrrolidin-1-yl)benzoic acid
Molecular Weight (g/mol):
191.23
Molecular Formula:
C11H13NO2
InChi Key:
HRVWGEKZONYEMK-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC(=CC=C1)N1CCCC1
1
–
2
of
2
results
2
3-Pyrrolidin-1-ylbenzoic acid, 97%, Thermo Scientific™
CAS: 72548-79-9 Molecular Formula: C11H13NO2 Molecular Weight (g/mol): 191.23 MDL Number: MFCD03426724 InChI Key: HRVWGEKZONYEMK-UHFFFAOYSA-N PubChem CID: 651721 IUPAC Name: 3-pyrrolidin-1-ylbenzoic acid SMILES: C1CCN(C1)C2=CC=CC(=C2)C(=O)O