missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrrolidin-1-ylbenzoic acid, 97%, Thermo Scientific™

Catalog No. CC38501DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC38501DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC38501DA Supplier Thermo Scientific Chemicals Supplier No. CC38501DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72548-79-9
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD03426724
InChI Key HRVWGEKZONYEMK-UHFFFAOYSA-N
PubChem CID 651721
IUPAC Name 3-pyrrolidin-1-ylbenzoic acid
SMILES C1CCN(C1)C2=CC=CC(=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-pyrrolidin-1-yl benzoic acid, 3-pyrrolidin-1-yl-benzoic acid, 3-1-pyrrolidinyl benzoic acid, benzoic acid, 3-1-pyrrolidinyl, 3-pyrrolidinylbenzoic acid, pubchem19568, 3-pyrrolidinobenzoic acid, 3-1-pyrrolidinyl benzoicacid, 1-3-carboxyphenyl pyrrolidine, benzoic acid,3-1-pyrrolidinyl
Formula Weight 191.23
Percent Purity 97%
Chemical Name or Material 3-Pyrrolidin-1-ylbenzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.