accessibility menu, dialog, popup

UserName

NH2SSNH2

CAS RN 722-27-0

IUPAC Name: 4-[(4-aminophenyl)disulfanyl]aniline
Synonyms: 4-[(4-aminophenyl)disulfanyl]aniline
Molecular Weight (g/mol): 248.36
Molecular Formula: C12H12N2S2
InChi Key: MERLDGDYUMSLAY-UHFFFAOYSA-N
SMILES: NC1=CC=C(SSC2=CC=C(N)C=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results