missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Dithiodianiline 98.0+%, TCI America™

Catalog No. D02915G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D02915G 5 g
D029125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D02915G Supplier TCI America Supplier No. D02915G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 722-27-0
Molecular Formula C12H12N2S2
Molecular Weight (g/mol) 248.362
MDL Number MFCD00007882
InChI Key MERLDGDYUMSLAY-UHFFFAOYSA-N
Synonym 4,4'-dithiodianiline, 4-aminophenyl disulfide, benzenamine, 4,4'-dithiobis, 4,4'-dithiobisaniline, 4,4'-diaminodiphenyl disulfide, p,p'-dithiodianiline, bis p-aminophenyl disulfide, bis 4-aminophenyl disulfide, p,p'-dithiobisaniline, 4-4-aminophenyl disulfanyl aniline
PubChem CID 12875
IUPAC Name 4-[(4-aminophenyl)disulfanyl]aniline
SMILES C1=CC(=CC=C1N)SSC2=CC=C(C=C2)N

Specifications

Melting Point 75°C
Quantity 5 g
Formula Weight 248.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dithiodianiline
EINECSNumber (3)-1127
RTECSNumber BX9580000
TSCA Yes