accessibility menu, dialog, popup

UserName

CAS RN 72150-22-2

IUPAC Name: 4,10-dioxatricyclo[5.2.1.0²,⁶]dec-8-en-3-one
Synonyms: 4,10-dioxatricyclo[5.2.1.0²,⁶]dec-8-en-3-one
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: KIAQKKYOUVIGII-UHFFFAOYNA-N
SMILES: O=C1OCC2C3OC(C=C3)C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results