missing translation for 'onlineSavingsMsg'
Learn More

4,10-Dioxatricyclo[5.2.1.0(2,6)]decan-8-en-3-one 97.0+%, TCI America™

Catalog No. D35525G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35525G 5 g
D35521G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D35525G Supplier TCI America Supplier No. D35525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 72150-22-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
InChI Key KIAQKKYOUVIGII-UHFFFAOYSA-N
Synonym 4,7-Epoxy-3a,4,7,7a-tetrahydroisobenzofuran-1(3H)-one
PubChem CID 331417
SMILES C1C2C3C=CC(C2C(=O)O1)O3

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,10-Dioxatricyclo[5.2.1.0(2,6)]decan-8-en-3-one