accessibility menu, dialog, popup

UserName

CAS RN 71963-77-4

H3CO(S)O(R)O(R)CH3(S)(R)CH3(S)(R)H3C(R)OO

CAS RN 71963-77-4

IUPAC Name: (1R,4S,5R,8S,9R,10S,12R,13R)-10-methoxy-1,5,9-trimethyl-11,14,15,16-tetraoxatetracyclo[10.3.1.0⁴,¹³.0⁸,¹³]hexadecane
Synonyms: artemether artemetero 蒿甲醚 β-artemether artemisininelactol methyl ether dihydroartemisinin methyl ether artemetherum dihydroquinghaosu methyl ether methyl-dihydroartemisinine
Molecular Weight (g/mol): 298.38
Molecular Formula: C16H26O5
InChi Key: SXYIRMFQILZOAM-HVNFFKDJSA-N
SMILES: CO[C@H]1O[C@@H]2O[C@@]3(C)CC[C@H]4[C@H](C)CC[C@@H]([C@H]1C)[C@@]24OO3
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)