missing translation for 'onlineSavingsMsg'
Learn More

Artemether 98.0+%, TCI America™

Catalog No. A21905G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21905G 5 g
A219025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21905G Supplier TCI America Supplier No. A21905G
Only null left

Chemical Identifiers

CAS 71963-77-4
Molecular Formula C16H26O5
Molecular Weight (g/mol) 298.379
MDL Number MFCD00866205
InChI Key SXYIRMFQILZOAM-ZSLLRAOYSA-N
PubChem CID 132451910
SMILES CC1CCC2C(C(OC3C24C1CCC(O3)(OO4)C)OC)C

Specifications

Melting Point 89°C
Color White
Quantity 5 g
Formula Weight 298.38
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Artemether
RTECSNumber KD4165000
TSCA No