accessibility menu, dialog, popup

UserName

NH2ClO

CAS RN 719-59-5

IUPAC Name: 2-benzoyl-4-chloroaniline
Synonyms: 2-amino-5-chlorobenzophenone
Molecular Weight (g/mol): 231.68
Molecular Formula: C13H10ClNO
InChi Key: ZUWXHHBROGLWNH-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (3)
  • (5)
  • (3)