missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzophenone 98.0+%, TCI America™

Catalog No. A0079500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0079500G 500 g
A007925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0079500G Supplier TCI America Supplier No. A0079500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 719-59-5
Molecular Formula C13H10ClNO
Molecular Weight (g/mol) 231.68
MDL Number MFCD00007839
InChI Key ZUWXHHBROGLWNH-UHFFFAOYSA-N
Synonym 2-amino-5-chlorobenzophenone, 2-amino-5-chlorophenyl phenyl methanone, 2-benzoyl-4-chloroaniline, methanone, 2-amino-5-chlorophenyl phenyl, 2-amino-5-chlorbenzophenone, 5-chloro-2-aminobenzophenone, 2-amino-5-chlorobenzylphenone, benzophenone, 2-amino-5-chloro, unii-fr80014zbt, 2-amino-5-chlorophenyl phenylmethanone
PubChem CID 12870
IUPAC Name 2-benzoyl-4-chloroaniline
SMILES NC1=CC=C(Cl)C=C1C(=O)C1=CC=CC=C1

Specifications

Melting Point 98°C
Color Yellow
Quantity 500 g
Formula Weight 231.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-chlorobenzophenone
EINECSNumber (9)-0075
RTECSNumber PC4933500
TSCA Yes