accessibility menu, dialog, popup

UserName

CAS RN 71617-20-4

IUPAC Name: hydrogen 1-benzyl-4-hydroxypiperidine-4-carbonitrile chloride
Synonyms: hydrogen 1-benzyl-4-hydroxypiperidine-4-carbonitrile chloride
Molecular Weight (g/mol): 252.74
Molecular Formula: C13H17ClN2O
InChi Key: BFCODNKBFLYVOU-UHFFFAOYSA-N
SMILES: [H+].[Cl-].OC1(CCN(CC2=CC=CC=C2)CC1)C#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results