accessibility menu, dialog, popup

UserName

CAS RN 7149-75-9

IUPAC Name: 4-chloro-3-methylaniline
Synonyms: 3-methyl-4-chloroaniline
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: HIHCTGNZNHSZPP-UHFFFAOYSA-N
SMILES: CC1=CC(N)=CC=C1Cl
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)