missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-methylaniline 98.0+%, TCI America™

Catalog No. A16105G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A16105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A16105G Supplier TCI America Supplier No. A16105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7149-75-9
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00066332
InChI Key HIHCTGNZNHSZPP-UHFFFAOYSA-N
Synonym 5-amino-2-chlorotoluene, benzenamine, 4-chloro-3-methyl, m-toluidine, 4-chloro, 3-methyl-4-chloroaniline, 4-chloro-m-toluidine, unii-6q2w6l7bwz, 6q2w6l7bwz, 4-chloro-3-methylphenylamine, 4-chloro-3-methyl-phenylamine, 4-chloro-3-methylphenyl amine
PubChem CID 23536
IUPAC Name 4-chloro-3-methylaniline
SMILES CC1=C(C=CC(=C1)N)Cl

Specifications

Melting Point 86°C
Boiling Point 241°C
Quantity 5 g
UN Number 2239
Formula Weight 141.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Chloro-3-methylaniline
EINECSNumber (3)-0265
TSCA No