CAS RN 7149-70-4
CAS RN 7149-70-4
IUPAC Name:
1-bromo-2-methyl-4-nitrobenzene
Synonyms:
1-bromo-2-methyl-4-nitrobenzene
Molecular Weight (g/mol):
216.03
Molecular Formula:
C7H6BrNO2
InChi Key:
HIMGPQVBNICCGL-UHFFFAOYSA-N
SMILES:
CC1=CC(=CC=C1Br)[N+]([O-])=O
1
–
1
of
1
results
1
2-Bromo-5-nitrotoluene 98.0+%, TCI America™
CAS: 7149-70-4 Molecular Formula: C7H6BrNO2 Molecular Weight (g/mol): 216.034 MDL Number: MFCD00007281 InChI Key: HIMGPQVBNICCGL-UHFFFAOYSA-N Synonym: 2-bromo-5-nitrotoluene,4-bromo-3-methylnitrobenzene,benzene, 1-bromo-2-methyl-4-nitro,bromo 5--2-nitrotoluene,2bromo-5-nitrotoluene,pubchem12553,acmc-209oiv,toluene, 2-bromo-5-nitro,ksc498a8d,methyl 2-bromo-5-nitrobenzene PubChem CID: 251672 IUPAC Name: 1-bromo-2-methyl-4-nitrobenzene SMILES: CC1=C(C=CC(=C1)[N+](=O)[O-])Br