missing translation for 'onlineSavingsMsg'
Learn More

Moclobemide 98.0+%, TCI America™

Catalog No. M273310MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M273310MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M273310MG Supplier TCI America Supplier No. M273310MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71320-77-9
Molecular Formula C13H17ClN2O2
Molecular Weight (g/mol) 268.741
MDL Number MFCD00865388
InChI Key YHXISWVBGDMDLQ-UHFFFAOYSA-N
Synonym moclobemide, aurorix, manerix, moclamine, moclobemid, moclobemida, moclobemidum, moclaime, moclamide, moclobamide
PubChem CID 4235
ChEBI CHEBI:83531
IUPAC Name 4-chloro-N-(2-morpholin-4-ylethyl)benzamide
SMILES C1COCCN1CCNC(=O)C2=CC=C(C=C2)Cl

Specifications

Melting Point 138°C
Color White
Quantity 10 mg
Formula Weight 268.74
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Moclobemide
RTECSNumber CV2462000
TSCA No