accessibility menu, dialog, popup

UserName

CAS RN 71217-46-4

OONCO

CAS RN 71217-46-4

IUPAC Name: 5-(isocyanatomethyl)-2H-1,3-benzodioxole
Synonyms: 5-(isocyanatomethyl)-2H-1,3-benzodioxole
Molecular Weight (g/mol): 177.16
Molecular Formula: C9H7NO3
InChi Key: RIUNOJGBBOBVDE-UHFFFAOYSA-N
SMILES: O=C=NCC1=CC2=C(OCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)