missing translation for 'onlineSavingsMsg'
Learn More

5-(Isocyanatomethyl)-1,3-benzodioxole, ≥97%, Thermo Scientific™

Catalog No. p-3175433
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC50806CB 250mg
CC50806DA 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC50806CB Supplier Thermo Scientific Chemicals Supplier No. CC50806CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 71217-46-4
Molecular Formula C9H7NO3
Molecular Weight (g/mol) 177.159
MDL Number MFCD08435909
InChI Key RIUNOJGBBOBVDE-UHFFFAOYSA-N
Synonym 5-isocyanatomethyl-2h-1,3-benzodioxole, 5-isocyanatomethyl-1,3-benzodioxole, 1,3-benzodioxole,5-isocyanatomethyl, 1,3-benzodioxole, 5-isocyanatomethyl, 3,4-methylenedioxy benzyl isocyanate, 1,3-benzodioxol-5-ylmethyl isocyanate, 5-isocyanatomethyl benzo d 1,3 dioxole, 2h-benzo 3,4-d 1,3-dioxolan-5-ylmethanisocyanate
PubChem CID 15932354
IUPAC Name 5-(isocyanatomethyl)-1,3-benzodioxole
SMILES C1OC2=C(O1)C=C(C=C2)CN=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 250mg
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 5-(Isocyanatomethyl)-1, 3-benzodioxole
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.