accessibility menu, dialog, popup

UserName

CH3(R)HO(S)HNOH2NOHO

CAS RN 7093-70-1

IUPAC Name: (2S,3R)-2-(2-aminoacetamido)-3-hydroxybutanoic acid
Synonyms: (2S,3R)-2-(2-aminoacetamido)-3-hydroxybutanoic acid
Molecular Weight (g/mol): 176.17
Molecular Formula: C6H12N2O4
InChi Key: OLIFSFOFKGKIRH-WUJLRWPWSA-N
SMILES: C[C@@H](O)[C@H](NC(=O)CN)C(O)=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results